ENA321765951 Enamine

2-(4-methylcyclohexylidene)acetic acid

  • MDL number MFCD00029539
  • Cas Number : 77842-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61...63
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77816

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77816
Availability details for
Quantity
  • Availability
  • Add to Cart