ENA321765952 Enamine

ethyl 2-(4-methanesulfonyl-2-nitrophenoxy)acetate

  • MDL number MFCD02976193
  • Cas Number : 431986-90-2
  • Molecular Weight : 303.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-78609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78609
Availability details for
Quantity
  • Availability
  • Add to Cart