ENA321765953 Enamine

2-(2,6-difluorophenyl)-2-(2,6-dimethylmorpholin-4-yl)ethan-1-amine

  • MDL number MFCD09732375
  • Cas Number : 1052562-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78611
Availability details for
Quantity
  • Availability
  • Add to Cart