ENA321765956 Enamine

3-(chloromethyl)-5-(2,3-dihydro-1,4-benzodioxin-2-yl)-1,2,4-oxadiazole

  • MDL number MFCD11189777
  • Cas Number : 1039860-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-78621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78621
Availability details for
Quantity
  • Availability
  • Add to Cart