ENA321765958 Enamine

4-(aminomethyl)-N,N,3-trimethylaniline

  • MDL number MFCD07786676
  • Cas Number : 57678-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78623
Availability details for
Quantity
  • Availability
  • Add to Cart