ENA321765963 Enamine

3,5-dimethyl-4-[(3-methylphenyl)methoxy]benzaldehyde

  • MDL number MFCD12558101
  • Cas Number : 1040033-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-78629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78629
Availability details for
Quantity
  • Availability
  • Add to Cart