ENA321765966 Enamine

1-(2,4-difluorobenzoyl)-1,4-diazepane

  • MDL number MFCD09043216
  • Cas Number : 926199-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-77818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77818
Availability details for
Quantity
  • Availability
  • Add to Cart