ENA321765968 Enamine

ethyl 2-(methylsulfanyl)propanoate

  • MDL number MFCD16089611
  • Cas Number : 40800-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78645
Availability details for
Quantity
  • Availability
  • Add to Cart