ENA321765978 Enamine

ethyl 2-chloropentanoate

  • MDL number MFCD16694312
  • Cas Number : 118688-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-77825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77825
Availability details for
Quantity
  • Availability
  • Add to Cart