ENA321765980 Enamine

5-amino-1-(4-fluorophenyl)-2,3-dihydro-1H-pyrazol-3-one

  • MDL number MFCD18917364
  • Cas Number : 1247169-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78663
Availability details for
Quantity
  • Availability
  • Add to Cart