ENA321765983 Enamine

(2-chlorophenyl)(pyridin-3-yl)methanamine dihydrochloride

  • MDL number MFCD08544232
  • Cas Number : 1311314-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 288...290
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78667

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78667
Availability details for
Quantity
  • Availability
  • Add to Cart