ENA321765984 Enamine

2-(3-methyl-2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoic acid

  • MDL number MFCD11643027
  • Cas Number : 553636-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78668
Availability details for
Quantity
  • Availability
  • Add to Cart