ENA321765985 Enamine

3-hydroxy-N-(2-hydroxyethyl)-N-methylbenzamide

  • MDL number MFCD12771085
  • Cas Number : 1183649-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77826

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77826
Availability details for
Quantity
  • Availability
  • Add to Cart