ENA321765987 Enamine

5-amino-N-[(4-methylphenyl)methyl]-1H-pyrazole-4-carboxamide

  • MDL number MFCD11210152
  • Cas Number : 1187064-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78672
Availability details for
Quantity
  • Availability
  • Add to Cart