ENA321765990 Enamine

1-(aminomethyl)cyclopentane-1-carboxamide

  • MDL number MFCD16118413
  • Cas Number : 1027337-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78709
Availability details for
Quantity
  • Availability
  • Add to Cart