ENA321765996 Enamine

2-(dibenzylamino)acetamide

  • MDL number MFCD06080632
  • Cas Number : 861056-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78715
Availability details for
Quantity
  • Availability
  • Add to Cart