ENA321766004 Enamine

3-(oxan-4-yl)propanoic acid

  • MDL number MFCD11226123
  • Cas Number : 40500-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78724

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-78724
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart