ENA321766005 Enamine

2-(4-bromophenyl)-2-(piperazin-1-yl)ethan-1-ol

  • MDL number MFCD12826667
  • Cas Number : 1183532-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78725
Availability details for
Quantity
  • Availability
  • Add to Cart