ENA321766016 Enamine

4-(methoxymethyl)-1,3-thiazol-2-amine

  • MDL number MFCD11166443
  • Cas Number : 640768-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78760

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-78760
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart