ENA321766024 Enamine

3,5-dimethyl-4-[(2-methylphenyl)methoxy]benzaldehyde

  • MDL number MFCD12461641
  • Cas Number : 1039819-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78774
Availability details for
Quantity
  • Availability
  • Add to Cart