ENA321766027 Enamine

2-amino-3,6-dibromobenzoic acid

  • MDL number MFCD01851371
  • Cas Number : 20776-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77747
Availability details for
Quantity
  • Availability
  • Add to Cart