ENA321766031 Enamine

4-ethyl-2-nitrophenol

  • MDL number MFCD16742721
  • Cas Number : 56520-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78784

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78784
Availability details for
Quantity
  • Availability
  • Add to Cart