ENA321766038 Enamine

2-(4-methylphenyl)-1-(pyridin-3-yl)ethan-1-amine

  • MDL number MFCD12153931
  • Cas Number : 1178359-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78797

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78797
Availability details for
Quantity
  • Availability
  • Add to Cart