ENA321766040 Enamine

2-[(2-hydroxyethyl)sulfanyl]benzoic acid

  • MDL number MFCD00020273
  • Cas Number : 58980-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125...127
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78802

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78802
Availability details for
Quantity
  • Availability
  • Add to Cart