ENA321766044 Enamine

ethyl 3-sulfanylbutanoate

  • MDL number MFCD07437968
  • Cas Number : 156472-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78807
Availability details for
Quantity
  • Availability
  • Add to Cart