ENA321766047 Enamine

2-(4-amino-2-chlorophenoxy)-N,N-dimethylacetamide

  • MDL number MFCD09739685
  • Cas Number : 169286-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78820

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78820
Availability details for
Quantity
  • Availability
  • Add to Cart