ENA321766055 Enamine

(4-methyl-1,2,5-oxadiazol-3-yl)methanol

  • MDL number MFCD00456449
  • Cas Number : 78441-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78848
Availability details for
Quantity
  • Availability
  • Add to Cart