ENA321766056 Enamine

N-ethyl-4-hydroxybenzamide

  • MDL number MFCD11130754
  • Cas Number : 27522-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78919
Availability details for
Quantity
  • Availability
  • Add to Cart