ENA321766066 Enamine

[2-(3,4-dimethoxyphenyl)ethyl](2-methylpropyl)amine

  • MDL number MFCD11141373
  • Cas Number : 745007-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78933
Availability details for
Quantity
  • Availability
  • Add to Cart