ENA321766071 Enamine

3-methyl-octahydro-1H-indole

  • MDL number MFCD02066175
  • Cas Number : 37865-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78030

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78030
Availability details for
Quantity
  • Availability
  • Add to Cart