ENA321766075 Enamine

2-(pyridin-2-yloxy)aniline

  • MDL number MFCD11650924
  • Cas Number : 74864-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78954
Availability details for
Quantity
  • Availability
  • Add to Cart