ENA321766077 Enamine

4-chloro-3-(methanesulfinylmethyl)aniline

  • MDL number MFCD14613631
  • Cas Number : 1250417-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78966

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78966
Availability details for
Quantity
  • Availability
  • Add to Cart