ENA321766079 Enamine

2,3-difluoro-5-nitrobenzoic acid

  • MDL number MFCD09859890
  • Cas Number : 942035-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78968

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78968
Availability details for
Quantity
  • Availability
  • Add to Cart