ENA321766081 Enamine

3-[(benzenesulfinyl)methyl]aniline

  • MDL number MFCD14650758
  • Cas Number : 1250005-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-78970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78970
Availability details for
Quantity
  • Availability
  • Add to Cart