ENA321766091 Enamine

1-ethyl-2-methylpiperidin-4-amine

  • MDL number MFCD14692737
  • Cas Number : 92351-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78981

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78981
Availability details for
Quantity
  • Availability
  • Add to Cart