ENA321766095 Enamine

4-(3-methylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02664016
  • Cas Number : 5330-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78987
Availability details for
Quantity
  • Availability
  • Add to Cart