ENA321766098 Enamine

(4-bromophenyl)(cyclopropyl)methanol

  • MDL number MFCD00019251
  • Cas Number : 70289-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78993
Availability details for
Quantity
  • Availability
  • Add to Cart