ENA321766103 Enamine

ethyl[1-(pyridin-3-yl)ethyl]amine

  • MDL number MFCD09941689
  • Cas Number : 5746-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78999

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-78999
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart