ENA321766114 Enamine

2-amino-3-chloro-5-methylbenzoic acid

  • MDL number MFCD11193638
  • Cas Number : 66490-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79026
Availability details for
Quantity
  • Availability
  • Add to Cart