ENA321766115 Enamine

N-(5-amino-2-methoxyphenyl)cyclohexanecarboxamide

  • MDL number MFCD09046332
  • Cas Number : 859179-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79027
Availability details for
Quantity
  • Availability
  • Add to Cart