ENA321766122 Enamine

3-[(oxan-4-ylsulfanyl)methyl]aniline

  • MDL number MFCD17268422
  • Cas Number : 1311318-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79034
Availability details for
Quantity
  • Availability
  • Add to Cart