ENA321766125 Enamine

2-(3-chlorophenyl)cyclopentan-1-ol

  • MDL number MFCD16818430
  • Cas Number : 91552-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79042
Availability details for
Quantity
  • Availability
  • Add to Cart