ENA321766127 Enamine

1-methoxy-4-methylpentan-3-one

  • MDL number MFCD14661410
  • Cas Number : 1250407-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79044
Availability details for
Quantity
  • Availability
  • Add to Cart