ENA321766129 Enamine

ethyl 3-hydroxyoctanoate

  • MDL number MFCD00082952
  • Cas Number : 7367-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79047

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79047
Availability details for
Quantity
  • Availability
  • Add to Cart