ENA321766137 Enamine

ethyl 5-(chlorosulfonyl)-2-methyl-3-nitrobenzoate

  • MDL number MFCD12464234
  • Cas Number : 1154155-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79065
Availability details for
Quantity
  • Availability
  • Add to Cart