ENA321766138 Enamine

2,2,5-trimethylhexan-3-one

  • MDL number MFCD12153388
  • Cas Number : 14705-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79066
Availability details for
Quantity
  • Availability
  • Add to Cart