ENA321766139 Enamine

3-[(tert-butylsulfanyl)methyl]aniline

  • MDL number MFCD11136243
  • Cas Number : 28288-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79067
Availability details for
Quantity
  • Availability
  • Add to Cart