ENA321766141 Enamine

2-[(3-methylcyclohexyl)oxy]propanoic acid

  • MDL number MFCD10690415
  • Cas Number : 1218150-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79069
Availability details for
Quantity
  • Availability
  • Add to Cart