ENA321766144 Enamine

2-chloro-6-(2,3,6-trimethylphenoxy)benzaldehyde

  • MDL number MFCD12542811
  • Cas Number : 1094477-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79075

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79075
Availability details for
Quantity
  • Availability
  • Add to Cart