ENA321766151 Enamine

4-methanesulfonylbenzene-1-sulfonamide

  • MDL number MFCD01657929
  • Cas Number : 667-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79083
Availability details for
Quantity
  • Availability
  • Add to Cart